MAT047022107 Matrix Scientific

3-Chlorobenzoyl isothiocyanate

  • MDL number MFCD00041103
  • Cas Number : 66090-36-6
  • Molecular Weight : 197.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Freezer
Country of Origin USA
Product Link http://www.matrixscientific.com/003318.html

Safety Information

hcodes Corrosive/Irritant/Store in Freezer
MSDS http://www.matrixscientific.com/003318.html
Availability details for
Quantity
  • Availability
  • Add to Cart