MAT047022124 Matrix Scientific

2-Fluorobenzyl isothiocyanate

  • MDL number MFCD00046817
  • Cas Number : 64382-80-5
  • Molecular Weight : 167.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Density 1.22
Country of Origin USA
Boiling point 130�mm
Product Link http://www.matrixscientific.com/003337.html

Safety Information

hcodes Irritant-Harmful/Refrigerate/Lachrymator
MSDS http://www.matrixscientific.com/003337.html
Availability details for
Quantity
  • Availability
  • Add to Cart