MAT047022150 Matrix Scientific

3,3,4,4,5,5,5-Heptafluoropentan-2-ol

  • MDL number MFCD00039619
  • Cas Number : 375-14-4
  • Molecular Weight : 214.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Density 1.483
Country of Origin USA
Boiling point 101-102
Product Link http://www.matrixscientific.com/001257.html

Safety Information

hcodes Irritant/Stored under Argon
MSDS http://www.matrixscientific.com/001257.html
Availability details for
Quantity
  • Availability
  • Add to Cart