MAT047022160 Matrix Scientific

6-(Bromomethyl)-4-chloro-2-(trifluoromethyl)-quinoline

  • MDL number MFCD00153079
  • Cas Number : 123637-51-4
  • Molecular Weight : 324.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 118-124
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/003388.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/003388.html
Availability details for
Quantity
  • Availability
  • Add to Cart