MAT047022161 Matrix Scientific

1,1,1,3,5,5,5-Heptafluoropentan-2,4-dione

  • MDL number MFCD00153187
  • Cas Number : 77968-17-3
  • Molecular Weight : 226.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Ambient
Country of Origin USA
Boiling point 70-72
Product Link http://www.matrixscientific.com/001258.html

Safety Information

hcodes Corrosive/Refrigerate
MSDS http://www.matrixscientific.com/001258.html
Availability details for
Quantity
  • Availability
  • Add to Cart