MAT047022165 Matrix Scientific

Ethyl 3-methyl-4,4,4-trifluorobutyrate

  • MDL number MFCD00040847
  • Cas Number : 6975-13-9
  • Molecular Weight : 184.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Boiling point 135-136
Product Link http://www.matrixscientific.com/003396.html

Safety Information

hcodes Flammable
MSDS http://www.matrixscientific.com/003396.html
Availability details for
Quantity
  • Availability
  • Add to Cart