MAT047022202 Matrix Scientific

4,5,6,7-Tetrafluoroindole

  • MDL number MFCD00236717
  • Cas Number : 16264-67-8
  • Molecular Weight : 189.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 90-93
Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/003442.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/003442.html
Availability details for
Quantity
  • Availability
  • Add to Cart