MAT047022218 Matrix Scientific

5,6,7,8-Tetrafluorobenzo-1,4-dioxane

  • MDL number MFCD00229993
  • Cas Number : 1743-87-9
  • Molecular Weight : 208.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 78-79
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/003461.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/003461.html
Availability details for
Quantity
  • Availability
  • Add to Cart