MAT047022223 Matrix Scientific

2,3,5,6-Tetrafluoro-4-(trifluoromethyl)phenyl-hydrazine

  • MDL number MFCD00012368
  • Cas Number : 1868-85-5
  • Molecular Weight : 248.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 84
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/003469.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/003469.html
Availability details for
Quantity
  • Availability
  • Add to Cart