MAT047022229 Matrix Scientific

2,3,5,6-Tetrafluorophenyl isothiocyanate

  • MDL number MFCD00041043
  • Cas Number : 207981-49-5
  • Molecular Weight : 207.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Boiling point 98�mm
Product Link http://www.matrixscientific.com/003475.html

Safety Information

hcodes Toxic/Refrigerate/Lachrymator
MSDS http://www.matrixscientific.com/003475.html
Availability details for
Quantity
  • Availability
  • Add to Cart