MAT047022234 Matrix Scientific

2,2,3,3-Tetrafluoropropyl p-toluenesulfonate

  • MDL number MFCD00155960
  • Cas Number : 786-31-2
  • Molecular Weight : 286.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 14-16
Purity 97
Density 1.397
Country of Origin USA
Boiling point 124�Torr
Product Link http://www.matrixscientific.com/003479.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/003479.html
Availability details for
Quantity
  • Availability
  • Add to Cart