MAT047022236 Matrix Scientific

3-Chlorotetrafluoropropionic acid

  • MDL number MFCD00153099
  • Cas Number : 661-82-5
  • Molecular Weight : 180.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Density 1.63
Country of Origin USA
Boiling point 130-131
Product Link http://www.matrixscientific.com/003481.html

Safety Information

hcodes Corrosive
MSDS http://www.matrixscientific.com/003481.html
Availability details for
Quantity
  • Availability
  • Add to Cart