MAT047022257 Matrix Scientific

1,1,2,2-Tetrafluoro-3-methoxypropane

  • MDL number MFCD01075284
  • Cas Number : 60598-17-6
  • Molecular Weight : 146.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Boiling point 64-68
Product Link http://www.matrixscientific.com/003500.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/003500.html
Availability details for
Quantity
  • Availability
  • Add to Cart