MAT047022269 Matrix Scientific

1,1,1,3-Tetrafluoro-2-(trifluoromethyl)-4-oxapent-2-ene

  • MDL number MFCD00054679
  • Cas Number : 360-53-2
  • Molecular Weight : 212.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Density 1.5078/20
Storage temp. Ambient
Country of Origin USA
Boiling point 98-104
Product Link http://www.matrixscientific.com/003511.html

Safety Information

hcodes Flammable/Toxic
MSDS http://www.matrixscientific.com/003511.html
Availability details for
Quantity
  • Availability
  • Add to Cart