MAT047022327 Matrix Scientific

1H,1H-Perfluoro-n-decyl acrylate

  • MDL number MFCD02183510
  • Cas Number : 335-83-1
  • Molecular Weight : 554.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Density 1.689/20
Storage temp. Refrigerate
Country of Origin USA
Boiling point 220�0 Torr
Product Link http://www.matrixscientific.com/003596.html

Safety Information

hcodes Irritant/Refrigerate
MSDS http://www.matrixscientific.com/003596.html
Availability details for
Quantity
  • Availability
  • Add to Cart