MAT047022328 Matrix Scientific

1H,1H-Perfluoro-n-decyl methacrylate

  • MDL number MFCD02183511
  • Cas Number : 23069-32-1
  • Molecular Weight : 568.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Boiling point 77�mHg
Product Link http://www.matrixscientific.com/003597.html

Safety Information

hcodes Irritant/Refrigerate/Lachrymator
MSDS http://www.matrixscientific.com/003597.html
Availability details for
Quantity
  • Availability
  • Add to Cart