MAT047022333 Matrix Scientific

4-Amino-2,5-difluorobenzonitrile

  • MDL number MFCD00190102
  • Cas Number : 112279-61-5
  • Molecular Weight : 154.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 95-97
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/001279.html

Safety Information

hcodes Toxic/Irritant
MSDS http://www.matrixscientific.com/001279.html
Availability details for
Quantity
  • Availability
  • Add to Cart