MAT047022361 Matrix Scientific

2,3,4-Trifluorobenzamide

  • MDL number MFCD00061265
  • Cas Number : 207919-09-3
  • Molecular Weight : 175.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 134-135
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/003631.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/003631.html
Availability details for
Quantity
  • Availability
  • Add to Cart