MAT047022362 Matrix Scientific

2,4,5-Trifluorobenzamide

  • MDL number MFCD00082478
  • Cas Number : 98349-23-6
  • Molecular Weight : 175.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/003632.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/003632.html
Availability details for
Quantity
  • Availability
  • Add to Cart