MAT047022371 Matrix Scientific

2,4,6-Trifluorobenzoyl chloride

  • MDL number MFCD00070810
  • Cas Number : 79538-29-7
  • Molecular Weight : 194.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Density 1.499
Country of Origin USA
Boiling point 167-168
Product Link http://www.matrixscientific.com/003640.html

Safety Information

hcodes Corrosive
MSDS http://www.matrixscientific.com/003640.html
Availability details for
Quantity
  • Availability
  • Add to Cart