MAT047022381 Matrix Scientific

2,4,5-Trifluorobenzyl bromide

  • MDL number MFCD00061209
  • Cas Number : 157911-56-3
  • Molecular Weight : 225.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/003649.html

Safety Information

hcodes Corrosive
MSDS http://www.matrixscientific.com/003649.html
Availability details for
Quantity
  • Availability
  • Add to Cart