MAT047022389 Matrix Scientific

2,3,5-Trifluorobenzyl alcohol

  • MDL number MFCD00083542
  • Cas Number : 67640-33-9
  • Molecular Weight : 162.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/003656.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/003656.html
Availability details for
Quantity
  • Availability
  • Add to Cart