MAT047022395 Matrix Scientific

2,3,5-Trifluorobenzyl bromide

  • MDL number MFCD00083543
  • Cas Number : 226717-83-5
  • Molecular Weight : 225.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/003662.html

Safety Information

hcodes Corrosive/Refrigerate/Lachrymator
MSDS http://www.matrixscientific.com/003662.html
Availability details for
Quantity
  • Availability
  • Add to Cart