MAT047022396 Matrix Scientific

2,4,5-Trifluorobenzyl chloride

  • MDL number MFCD00236319
  • Cas Number : 243139-71-1
  • Molecular Weight : 180.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Density 1.689
Country of Origin USA
Product Link http://www.matrixscientific.com/003663.html

Safety Information

hcodes Corrosive
MSDS http://www.matrixscientific.com/003663.html
Availability details for
Quantity
  • Availability
  • Add to Cart