MAT047022398 Matrix Scientific

2,4,5-Trifluorobenzenesulfonyl chloride

  • MDL number MFCD00077600
  • Cas Number : 220227-21-4
  • Molecular Weight : 230.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Density 1.657
Country of Origin USA
Product Link http://www.matrixscientific.com/003665.html

Safety Information

hcodes Corrosive/Irritant/Refrigerate
MSDS http://www.matrixscientific.com/003665.html
Availability details for
Quantity
  • Availability
  • Add to Cart