MAT047022400 Matrix Scientific

4-Bromo-3,5-difluoroaniline

  • MDL number MFCD00236593
  • Cas Number : 203302-95-8
  • Molecular Weight : 208.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 99-101
Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/001285.html

Safety Information

hcodes Toxic
MSDS http://www.matrixscientific.com/001285.html
Availability details for
Quantity
  • Availability
  • Add to Cart