MAT047022401 Matrix Scientific

2,4,5-Trifluorobenzenesulfonamide

  • MDL number MFCD01569462
  • Cas Number : 287172-63-8
  • Molecular Weight : 211.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 118-120
Country of Origin USA
Product Link http://www.matrixscientific.com/003666.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/003666.html
Availability details for
Quantity
  • Availability
  • Add to Cart