MAT047022405 Matrix Scientific

2,3,4-Trifluorobenzonitrile

  • MDL number MFCD00013288
  • Cas Number : 143879-80-5
  • Molecular Weight : 157.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Density 1.12
Country of Origin USA
Boiling point 116-118�0mm
Product Link http://www.matrixscientific.com/003670.html

Safety Information

hcodes Toxic/Irritant
MSDS http://www.matrixscientific.com/003670.html
Availability details for
Quantity
  • Availability
  • Add to Cart