MAT047022419 Matrix Scientific

4,4,4-Trifluorobut-2-en-1-ol

  • MDL number MFCD00077603
  • Cas Number : 674-53-3
  • Molecular Weight : 126.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Boiling point 118.5�0mm
Product Link http://www.matrixscientific.com/003683.html

Safety Information

hcodes Flammable
MSDS http://www.matrixscientific.com/003683.html
Availability details for
Quantity
  • Availability
  • Add to Cart