MAT047022426 Matrix Scientific

2,3,6-Trifluorocinnamic acid

  • MDL number MFCD00061213
  • Cas Number : 230295-12-2
  • Molecular Weight : 202.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/003690.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/003690.html
Availability details for
Quantity
  • Availability
  • Add to Cart