MAT047022466 Matrix Scientific

2-Bromo-4,5-dimethoxyphenylacetonitrile

  • MDL number MFCD00016385
  • Cas Number : 51655-39-1
  • Molecular Weight : 256.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 90-91
Country of Origin USA
Product Link http://www.matrixscientific.com/001291.html

Safety Information

hcodes Irritant/Irritant-Harmful
MSDS http://www.matrixscientific.com/001291.html
Availability details for
Quantity
  • Availability
  • Add to Cart