MAT047022474 Matrix Scientific

N-[4-(Trifluoromethyl)phenyl]-4-fluorobenzamide

  • MDL number MFCD00045094
  • Cas Number : 2054-01-5
  • Molecular Weight : 283.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 153-154
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/003747.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/003747.html
Availability details for
Quantity
  • Availability
  • Add to Cart