MAT047022478 Matrix Scientific

1-[3-(Trifluoromethyl)pyrid-2-yl]piperazine

  • MDL number MFCD00114705
  • Cas Number : 87394-63-6
  • Molecular Weight : 231.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 45-47
Storage temp. Ambient/argon
Country of Origin USA
Boiling point 106�5mm
Product Link http://www.matrixscientific.com/003752.html

Safety Information

hcodes Corrosive/Stored under Argon
MSDS http://www.matrixscientific.com/003752.html
Availability details for
Quantity
  • Availability
  • Add to Cart