MAT047022496 Matrix Scientific

Ethyl perfluoropentanoate

  • MDL number MFCD00077523
  • Cas Number : 424-36-2
  • Molecular Weight : 292.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Density 1.48
Storage temp. Ambient
Country of Origin USA
Boiling point 121
Product Link http://www.matrixscientific.com/003772.html

Safety Information

hcodes Flammable
MSDS http://www.matrixscientific.com/003772.html
Availability details for
Quantity
  • Availability
  • Add to Cart