MAT047022501 Matrix Scientific

1H,1H-Perfluoropentan-1-ol

  • MDL number MFCD00236714
  • Cas Number : 355-28-2
  • Molecular Weight : 250.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Density 1.664
Storage temp. Ambient
Country of Origin USA
Boiling point 77�3mm
Product Link http://www.matrixscientific.com/003776.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/003776.html
Availability details for
Quantity
  • Availability
  • Add to Cart