MAT047022511 Matrix Scientific

1,1,1,5,5,6,6,7,7,7-Decafluoro-2,4-heptanedione

  • MDL number MFCD00040961
  • Cas Number : 20583-66-8
  • Molecular Weight : 308.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Boiling point 98-100
Product Link http://www.matrixscientific.com/001295.html

Safety Information

hcodes Corrosive
MSDS http://www.matrixscientific.com/001295.html
Availability details for
Quantity
  • Availability
  • Add to Cart