MAT047022517 Matrix Scientific

3,4,5-Trifluorophenylacetic acid

  • MDL number MFCD00083532
  • Cas Number : 209991-62-8
  • Molecular Weight : 190.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98-101
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/003790.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/003790.html
Availability details for
Quantity
  • Availability
  • Add to Cart