MAT047022519 Matrix Scientific

2,3,5-Trifluorophenylacetonitrile

  • MDL number MFCD00083546
  • Cas Number : 243666-14-0
  • Molecular Weight : 171.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/003792.html

Safety Information

hcodes Toxic
MSDS http://www.matrixscientific.com/003792.html
Availability details for
Quantity
  • Availability
  • Add to Cart