MAT047022523 Matrix Scientific

2,4,6-Trifluorophenylacetonitrile

  • MDL number MFCD00061221
  • Cas Number : 220227-80-5
  • Molecular Weight : 171.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/003795.html

Safety Information

hcodes Toxic
MSDS http://www.matrixscientific.com/003795.html
Availability details for
Quantity
  • Availability
  • Add to Cart