MAT047022542 Matrix Scientific

2,4,6-Trifluoropyrimidine

  • MDL number MFCD00039705
  • Cas Number : 696-82-2
  • Molecular Weight : 134.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Ambient/argon
Country of Origin USA
Boiling point 98
Product Link http://www.matrixscientific.com/003818.html

Safety Information

hcodes Corrosive/Stored under Argon/Moisture Sensitive/Refrigerate
MSDS http://www.matrixscientific.com/003818.html
Availability details for
Quantity
  • Availability
  • Add to Cart