MAT047022545 Matrix Scientific

Methyl 6-fluorosalicylate

  • MDL number MFCD00153207
  • Cas Number : 72373-81-0
  • Molecular Weight : 170.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Boiling point 58-60�3mm
Product Link http://www.matrixscientific.com/003820.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/003820.html
Availability details for
Quantity
  • Availability
  • Add to Cart