MAT047022566 Matrix Scientific

2,6-Dichloro-4-(trifluoromethyl)nicotinonitrile

  • MDL number MFCD00084977
  • Cas Number : 13600-42-5
  • Molecular Weight : 241

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 39-40
Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/001302.html

Safety Information

hcodes Toxic
MSDS http://www.matrixscientific.com/001302.html
Availability details for
Quantity
  • Availability
  • Add to Cart