MAT047022596 Matrix Scientific

4-Ethoxyphenylacetonitrile

  • MDL number MFCD00016401
  • Cas Number : 6775-77-5
  • Molecular Weight : 161.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 44-47
Purity 98
Country of Origin USA
Boiling point 150-151� Torr
Product Link http://www.matrixscientific.com/003895.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/003895.html
Availability details for
Quantity
  • Availability
  • Add to Cart