MAT047022598 Matrix Scientific

2-Ethoxyphenylacetonitrile

  • MDL number MFCD00016390
  • Cas Number : 74205-51-9
  • Molecular Weight : 161.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 32-34
Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/003897.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/003897.html
Availability details for
Quantity
  • Availability
  • Add to Cart