MAT047022606 Matrix Scientific

4-Chlorophenylacetone

  • MDL number MFCD00045214
  • Cas Number : 5586-88-9
  • Molecular Weight : 168.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 6-8
Purity 97
Density 1.1397
Country of Origin USA
Boiling point 98�m
Product Link http://www.matrixscientific.com/003905.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/003905.html
Availability details for
Quantity
  • Availability
  • Add to Cart