MAT047022634 Matrix Scientific

Dimethyl hexadecafluorosebacate

  • MDL number MFCD00236604
  • Cas Number : 4590-24-3
  • Molecular Weight : 518.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Boiling point 144-146
Product Link http://www.matrixscientific.com/003936.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/003936.html
Availability details for
Quantity
  • Availability
  • Add to Cart