MAT047022637 Matrix Scientific

Dimethyl hexafluoroglutarate

  • MDL number MFCD00155851
  • Cas Number : 1513-62-8
  • Molecular Weight : 268.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point -33- -31
Purity 98
Density 1.4856/20
Country of Origin USA
Boiling point 100� Torr
Product Link http://www.matrixscientific.com/003940.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/003940.html
Availability details for
Quantity
  • Availability
  • Add to Cart