MAT047022649 Matrix Scientific

4-Chlorothioanisole

  • MDL number MFCD00013643
  • Cas Number : 123-09-1
  • Molecular Weight : 158.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 18-19
Purity 99
Density 1.22
Country of Origin USA
Boiling point 170
Product Link http://www.matrixscientific.com/003954.html

Safety Information

hcodes Stench
MSDS http://www.matrixscientific.com/003954.html
Availability details for
Quantity
  • Availability
  • Add to Cart