MAT047022685 Matrix Scientific

2,5-Dimethoxybenzhydrazide

  • MDL number MFCD00017049
  • Cas Number : 17894-25-6
  • Molecular Weight : 196.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 89-91
Country of Origin USA
Product Link http://www.matrixscientific.com/003996.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/003996.html
Availability details for
Quantity
  • Availability
  • Add to Cart