MAT047022687 Matrix Scientific

3-Amino-2-(2,4-difluorophenoxy)pyridine

  • MDL number MFCD00067802
  • Cas Number : 175135-63-4
  • Molecular Weight : 222.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 87-89
Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/001316.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/001316.html
Availability details for
Quantity
  • Availability
  • Add to Cart